2-Bromo-5-chlorothiophene ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B1996-25G B1996-5G
TCIAB1996-25GEA 253 EUR
TCIAB1996-25G TCIAB1996-5G
2-Bromo-5-chlorothiophene ≥95.0% (by GC)
2-Bromo-5-chlorothiophene
Formule: C₄H₂BrClS
Kookpunt: 45 - 46 °C (1 -rr)
Smeltpunt: –2 °C
Dichtheid: 1,803 g/cm³ (25 °C)
Vlampunt: 85°C
MDL Nummer: MFCD00014523
CAS nummer: 2873-18-9
EINECS: 220-708-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ef98e130106812d9c7b6a5d4ea85901cd1f5f13&vid=4ef98e130106812d9c7b6a5d4ea85901cd1f5f13&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %