Thiophene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T0223-25ML T0223-500ML
TCIAT0223-25MLEA 18.7 EUR
TCIAT0223-25ML TCIAT0223-500ML
Thiophene ≥98.0% (by GC)
Thiophene
Formule: C₄H₄S
Kookpunt: 84°C
Smeltpunt: –38°C
Dichtheid: 1,06 g/cm³ (20 °C)
Vlampunt: –1°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005413
CAS nummer: 110-02-1
EINECS: 203-729-4
UN: 2414
ADR: 3,II
Merck Index: 14,09353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cf585f00416deda1c555da762e733ee399fe29a&vid=1cf585f00416deda1c555da762e733ee399fe29a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %