2-Methylthiophene ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M0635-25ML M0635-500ML
TCIAM0635-25MLEA 75.9 EUR
TCIAM0635-25ML TCIAM0635-500ML
2-Methylthiophene ≥97.0% (by GC)
2-Methylthiophene
Formule: C₅H₆S
Kookpunt: 113°C
Smeltpunt: –63 °C
Dichtheid: 1,02 g/cm³ (20 °C)
Vlampunt: 8°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005451
CAS nummer: 554-14-3
EINECS: 209-063-0
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5924a4e99bd0ff08c135f2e8b0ad3ca86367edcf&vid=5924a4e99bd0ff08c135f2e8b0ad3ca86367edcf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR