3,4-(Methylenedioxy)aniline ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
5-Amino-1,3-benzodioxole
M0741-25G
M0741-100G
TCIAM0741-25GEA
67.1
EUR
TCIAM0741-25G
TCIAM0741-100G
3,4-(Methylenedioxy)aniline ≥98.0% (by GC, titration analysis)
3,4-(Methylenedioxy)aniline
Formule:
C₇H₇NO₂ MW: 137.14 g/mol Kookpunt: 156°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00005832 CAS nummer: 14268-66-7 EINECS: 238-161-6 |
|
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 41.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...