(±)-Methyl-2-chloropropionate ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C0970-25ML C0970-500ML C0970-100ML
TCIAC0970-25MLEA 18.7 EUR
TCIAC0970-25ML TCIAC0970-500ML TCIAC0970-100ML
(±)-Methyl-2-chloropropionate ≥95.0% (by GC)
(±)-Methyl-2-chloropropionate
Formule: C₄H₇ClO₂
Kookpunt: 132°C
Vlampunt: 38°C
MDL Nummer: MFCD00000863
CAS nummer: 17639-93-9
EINECS: 241-624-5
UN: 2933
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d85a1ce8bb87d03db3edbf2d0e9febde11e88441&vid=d85a1ce8bb87d03db3edbf2d0e9febde11e88441&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR