Citrazinic acid ≥96.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C0571-25G C0571-500G
TCIAC0571-25GEA 46.2 EUR
TCIAC0571-25G TCIAC0571-500G
Citrazinic acid ≥96.0% (by titrimetric analysis)
Citrazinic acid
Formule: C₆H₅NO₄
MDL Nummer: MFCD00006274
CAS nummer: 99-11-6
EINECS: 202-731-2
Merck Index: 14,02325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15c6ebbd9a9c740ca51ee3ff3f7baaffc69dfb0a&vid=15c6ebbd9a9c740ca51ee3ff3f7baaffc69dfb0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR