Chlorohydroquinone ≥85.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C0185-25G C0185-500G
TCIAC0185-25GEA 40.7 EUR
TCIAC0185-25G TCIAC0185-500G
Chlorohydroquinone ≥85.0% (by GC)
Chlorohydroquinone
Formule: C₆H₅ClO₂
Kookpunt: 263°C
MDL Nummer: MFCD00002341
CAS nummer: 615-67-8
EINECS: 210-442-8
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a2d441febbf682e13847f9fd1b5d0e0f4334462&vid=9a2d441febbf682e13847f9fd1b5d0e0f4334462&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 85.0 %
Purity (Cerium redox Method) min. 97.0 %
Melting point 98.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR