Undecanoyl chloride ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

U0026-10ML U0026-25ML
TCIAU0026-10MLEA 49.5 EUR
TCIAU0026-10ML TCIAU0026-25ML
Undecanoyl chloride ≥97.0% (by GC, titration analysis)
Undecanoyl chloride
Formule: C₁₁H₂₁ClO
Kookpunt: 130°C
MDL Nummer: MFCD00000739
CAS nummer: 17746-05-3
EINECS: 241-741-1
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1bf05feba7a47186c74ae90094170e6196777d4&vid=b1bf05feba7a47186c74ae90094170e6196777d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR