Butyl mercaptoacetate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Butyl Mercaptoacetate, Butyl Thioglycolate, Mercaptoacetic acid butyl ester, Thioglycolic acid butyl ester

T0903-25ML T0903-500ML
TCIAT0903-25MLEA 20.9 EUR
TCIAT0903-25ML TCIAT0903-500ML
Butyl mercaptoacetate ≥98.0% (by GC)
Butyl mercaptoacetate
Formule: C₆H₁₂O₂S
MW: 148.23 g/mol
Kookpunt: 194°C
Vlampunt: 66°C
MDL Nummer: MFCD00022087
CAS nummer: 10047-28-6
EINECS: 233-156-5
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2795db80c7dbe6e1d9045188880865fcb0e8f616&vid=2795db80c7dbe6e1d9045188880865fcb0e8f616&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR