Ethyl mercaptoacetate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Ethyl 2-mercaptoacetate, Ethyl thioglycolate

T0211-25G T0211-500G
TCIAT0211-25GEA 20.9 EUR
TCIAT0211-25G TCIAT0211-500G
Ethyl mercaptoacetate ≥97.0% (by GC)
Ethyl mercaptoacetate
Formule: C₄H₈O₂S
MW: 120.17 g/mol
Kookpunt: 157°C
Vlampunt: 48°C
MDL Nummer: MFCD00004874
CAS nummer: 623-51-8
EINECS: 210-800-3
UN: 1992
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b2cc666965712ca3942946d5c801631e7b1be5b&vid=0b2cc666965712ca3942946d5c801631e7b1be5b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR