Chloromethyl chloroformate ≥95.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C1396-25G C1396-100G
TCIAC1396-25GEA 191 EUR
TCIAC1396-25G TCIAC1396-100G
Chloromethyl chloroformate ≥95.0% (by GC, titration analysis)
Chloromethyl chloroformate
Formule: C₂H₂Cl₂O₂
Kookpunt: 108°C
Vlampunt: 95°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00077688
CAS nummer: 22128-62-7
EINECS: 244-793-3
UN: 2745
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e804568bab7903101cf4cc780b18b240b840005b&vid=e804568bab7903101cf4cc780b18b240b840005b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %