2-(Methylthio)ethanol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M0358-25ML
M0358-100ML
TCIAM0358-25MLEA
45.1
EUR
TCIAM0358-25ML
TCIAM0358-100ML
2-(Methylthio)ethanol ≥98.0% (by GC)
2-(Methylthio)ethanol
Formule:
C₃H₈OS Kookpunt: 81°C Vlampunt: 70°C |
MDL Nummer:
MFCD00002908 CAS nummer: 5271-38-5 EINECS: 226-090-3 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...