5-Decyne ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D1190-25ML D1190-5ML
TCIAD1190-25MLEA 242 EUR
TCIAD1190-25ML TCIAD1190-5ML
5-Decyne ≥98.0% (by GC)
5-Decyne
Formule: C₁₀H₁₈
Kookpunt: 177°C
Vlampunt: 54°C
MDL Nummer: MFCD00027277
CAS nummer: 1942-46-7
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46a8d496383d8047458b10fe430a2f1751cef12f&vid=46a8d496383d8047458b10fe430a2f1751cef12f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %