Norbornene ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

N0166-25G N0166-400G
TCIAN0166-25GEA 18.7 EUR
TCIAN0166-25G TCIAN0166-400G
Norbornene ≥99.0% (by GC)
Norbornene
Formule: C₇H₁₀
Kookpunt: 96°C
MDL Nummer: MFCD00082304
CAS nummer: 498-66-8
EINECS: 207-866-0
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5684240730ef49c4e8e7275c1220a4cb8cac910b&vid=5684240730ef49c4e8e7275c1220a4cb8cac910b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 45.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR