Phenylbutazone ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
P1686-25G
P1686-500G
TCIAP1686-25GEA
28.6
EUR
TCIAP1686-25G
TCIAP1686-500G
Phenylbutazone ≥98.0% (by HPLC, titration analysis)
Phenylbutazone
Formule:
C₁₉H₂₀N₂O₂ |
MDL Nummer:
MFCD00005500 CAS nummer: 50-33-9 EINECS: 200-029-0 UN: 2811 ADR: 6.1,III Merck Index: 14,07277 |
|
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 104.0 to 108.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...