5-Bromo-2-hydroxypyrimidine ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B2848-25G B2848-5G
TCIAB2848-25GEA 69.3 EUR
TCIAB2848-25G TCIAB2848-5G
5-Bromo-2-hydroxypyrimidine ≥95.0% (by GC)
5-Bromo-2-hydroxypyrimidine
Formule: C₄H₃BrN₂O
Smeltpunt: 235°C
MDL Nummer: MFCD01318956
CAS nummer: 38353-06-9
EINECS: 253-896-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ca430c5e1419c57c61510d0acc9886e43f69240&vid=4ca430c5e1419c57c61510d0acc9886e43f69240&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %