Methyl phenyl sulphoxide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (Methylsulphinyl)benzene

M1148-25G M1148-5G
TCIAM1148-25GEA 141 EUR
TCIAM1148-25G TCIAM1148-5G
Methyl phenyl sulphoxide ≥98.0% (by GC)
Methyl phenyl sulphoxide
Formule: C₇H₈OS
MDL Nummer: MFCD00002088
CAS nummer: 1193-82-4
EINECS: 214-781-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b482c615097ecd466bc84ecada80db330d08373&vid=5b482c615097ecd466bc84ecada80db330d08373&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR