DL-Panthenol ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Panthenol, DL-Pantothenol, 2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

P1318-25G P1318-500G
TCIAP1318-25GEA 18.7 EUR
TCIAP1318-25G TCIAP1318-500G
DL-Panthenol ≥98.0% (by total nitrogen basis)
DL-Panthenol
Formule: C₉H₁₉NO₄
MW: 205.25 g/mol
MDL Nummer: MFCD00002944
CAS nummer: 16485-10-2
EINECS: 240-540-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33ab78eab58d4ea6b992d636288d8ec3bdeeb141&vid=33ab78eab58d4ea6b992d636288d8ec3bdeeb141&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 65.0 to 69.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR