Triclosan ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 5-Chloro-2-(2,4-dichlorophenoxy)phenol, 5-Chloro-2-(2, 4-dichlorophenoxy) phenol

T1872-25G T1872-500G T1872-100G
TCIAT1872-25GEA 45.1 EUR
TCIAT1872-25G TCIAT1872-500G TCIAT1872-100G
Triclosan ≥98.0% (by GC)
Triclosan
Formule: C₁₂H₇Cl₃O₂
Kookpunt: 290°C
MDL Nummer: MFCD00800992
CAS nummer: 3380-34-5
EINECS: 222-182-2
UN: 3077
ADR: 9,III
Merck Index: 14,09657
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c0f79b9dd37651096ee61c55f704b75a51e6d0a&vid=8c0f79b9dd37651096ee61c55f704b75a51e6d0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR