2-Methyl-2-oxazoline ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2-Methyl-4,5-dihydrooxazole
M0857-25G M0857-500G M0857-100G
TCIAM0857-25GEA 33 EUR
TCIAM0857-25G TCIAM0857-500G TCIAM0857-100G
2-Methyl-2-oxazoline ≥98.0% (by GC)
2-Methyl-2-oxazoline
Formule: C₄H₇NO
MW: 85.11 g/mol
Kookpunt: 110°C
Vlampunt: 20°C
MDL Nummer: MFCD00005298
CAS nummer: 1120-64-5
EINECS: 214-316-3
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=960a844cdadc276e2c45100ec06e27fbe1d95293&vid=960a844cdadc276e2c45100ec06e27fbe1d95293&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %