Rhodanine-N-propionic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3-(4-Oxo-2-thioxothiazolidin-3-yl)propanoic acid

R0060-25G R0060-5G
TCIAR0060-25GEA 422 EUR
TCIAR0060-25G TCIAR0060-5G
Rhodanine-N-propionic acid ≥98.0% (by titrimetric analysis)
Rhodanine-N-propionic acid
Formule: C₆H₇NO₃S₂
MDL Nummer: MFCD00022558
CAS nummer: 7025-19-6
EINECS: 230-307-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67ac0a6118ae70fd6b411ad66ce096dbd500e84f&vid=67ac0a6118ae70fd6b411ad66ce096dbd500e84f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR