1,3-Diethynylbenzene ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D2496-1G D2496-5G
TCIAD2496-1GEA 45.1 EUR
TCIAD2496-1G TCIAD2496-5G
1,3-Diethynylbenzene ≥96.0% (by GC)
1,3-Diethynylbenzene
Formule: C₁₀H₆
Kookpunt: 78°C
Vlampunt: 64°C
MDL Nummer: MFCD00041686
CAS nummer: 1785-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64efa4c7be06d82a3ffef07dd784ce97bd64d6ff&vid=64efa4c7be06d82a3ffef07dd784ce97bd64d6ff&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 96.0 %