2,1,3-Benzothiadiazole ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B0921-10G
B0921-25G
TCIAB0921-10GEA
50.6
EUR
TCIAB0921-10G
TCIAB0921-25G
2,1,3-Benzothiadiazole ≥99.0% (by GC)
2,1,3-Benzothiadiazole
Formule:
C₆H₄N₂S Kookpunt: 64°C |
MDL Nummer:
MFCD00005809 CAS nummer: 273-13-2 EINECS: 205-985-2 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 43.0 to 47.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...