(1H,3H)-Quinazoline-2,4-dione ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B3381-25G
B3381-5G
TCIAB3381-25GEA
149
EUR
TCIAB3381-25G
TCIAB3381-5G
(1H,3H)-Quinazoline-2,4-dione ≥98.0% (by HPLC, titration analysis)
(1H,3H)-Quinazoline-2,4-dione
Formule:
C₈H₆N₂O₂ Smeltpunt: 354°C |
MDL Nummer:
MFCD00006699 CAS nummer: 86-96-4 EINECS: 201-712-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...