Purpurin(1,2,4-trihydroxy-9,10-anthraquinone)
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11752-5 11752-10 11752-25
CAYM11752-5EA 49.2 EUR
CAYM11752-5 CAYM11752-10 CAYM11752-25
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone)
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone)
Formule: C₁₄H₈O₅
MDL Nummer: MFCD00001203
CAS nummer: 81-54-9
EINECS: 201-359-8