Chlorpropham (isopropyl-3-chlorocarbanilate) ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Isopropyl N-(3-Chlorophenyl)carbamate, N-(3-Chlorophenyl)carbamic acid Isopropyl ester, Chlorpropham

C2555-1G C2555-25G
TCIAC2555-1GEA 27.5 EUR
TCIAC2555-1G TCIAC2555-25G
Chlorpropham (isopropyl-3-chlorocarbanilate) ≥99.0% (by GC)
Chlorpropham (isopropyl-3-chlorocarbanilate)
Formule: C₁₀H₁₂ClNO₂
MDL Nummer: MFCD00037108
CAS nummer: 101-21-3
EINECS: 202-925-7
UN: 3077
ADR: 9,III
Merck Index: 14,02187
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7759db50e4da7b6e1d5673863ecb610f9da3454a&vid=7759db50e4da7b6e1d5673863ecb610f9da3454a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 40.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR