4-Methoxyphenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M0637-25G M0637-5G
TCIAM0637-25GEA 91.3 EUR
TCIAM0637-25G TCIAM0637-5G
4-Methoxyphenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)
4-Methoxyphenylhydrazinium chloride
Formule: C₇H₁₁ClN₂O
MDL Nummer: MFCD00012945
CAS nummer: 19501-58-7
EINECS: 243-115-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8a633123ff096f0a1c88faa06bb260670282864&vid=b8a633123ff096f0a1c88faa06bb260670282864&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Purple powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Potassium iodate Method) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR