Tetraethylurea ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 1,1,3,3-Tetraethylurea
T0587-25ML T0587-100ML
TCIAT0587-25MLEA 73.7 EUR
TCIAT0587-25ML TCIAT0587-100ML
Tetraethylurea ≥99.0% (by GC)
Tetraethylurea
Formule: C₉H₂₀N₂O
MW: 172.27 g/mol
Kookpunt: 214°C
Vlampunt: 81°C
MDL Nummer: MFCD00042881
CAS nummer: 1187-03-7
EINECS: 214-696-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7d71f06cd0c9c715404b8ea79ccf51a4c0c9d66&vid=b7d71f06cd0c9c715404b8ea79ccf51a4c0c9d66&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %