Fluorene-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

F0541-1G F0541-5G
TCIAF0541-1GEA 40.7 EUR
TCIAF0541-1G TCIAF0541-5G
Fluorene-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Fluorene-1-carboxylic acid
Formule: C₁₄H₁₀O₂
Kookpunt: 100°C
MDL Nummer: MFCD00001130
CAS nummer: 6276-03-5
EINECS: 228-472-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30b35f6ad5b8fd8a45d8e8ae8c252815102599c6&vid=30b35f6ad5b8fd8a45d8e8ae8c252815102599c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR