Decylphosphonic acid ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
n-Decylphosphonic acid, 1-Decylphosphonic acid
D3884-5G
D3884-1G
TCIAD3884-5GEA
233
EUR
TCIAD3884-5G
TCIAD3884-1G
Decylphosphonic acid ≥98.0% (by titrimetric analysis)
Decylphosphonic acid
Formule:
C₁₀H₂₃O₃P MW: 222.26 g/mol |
MDL Nummer:
MFCD00015830 CAS nummer: 6874-60-8 EINECS: 229-975-2 UN: 3261 ADR: 8 III |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...