Octadecylphosphonic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: n-Octadecylphosphonic acid, ODPA

O0371-1G O0371-5G
TCIAO0371-1GEA 68.2 EUR
TCIAO0371-1G TCIAO0371-5G
Octadecylphosphonic acid ≥98.0% (by titrimetric analysis)
Octadecylphosphonic acid
Formule: C₁₈H₃₉O₃P
MDL Nummer: MFCD00015838
CAS nummer: 4724-47-4
EINECS: 225-216-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a711920221a5d07665875bb880547d04096d7c7&vid=0a711920221a5d07665875bb880547d04096d7c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR