Triethyl-3-phosphonopropionate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D1524-25G D1524-250G
TCIAD1524-25GEA 61.6 EUR
TCIAD1524-25G TCIAD1524-250G
Triethyl-3-phosphonopropionate ≥98.0% (by GC)
Triethyl-3-phosphonopropionate
Formule: C₉H₁₉O₅P
Kookpunt: 95°C
MDL Nummer: MFCD00015167
CAS nummer: 3699-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c24bb4af6cf7ad2dfe4e8e69d4678759e01f6734&vid=c24bb4af6cf7ad2dfe4e8e69d4678759e01f6734&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR