Triethyl methanetricarboxylate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

T1481-25G T1481-5G
TCIAT1481-25GEA 145 EUR
TCIAT1481-25G TCIAT1481-5G
Triethyl methanetricarboxylate ≥97.0% (by GC)
Triethyl methanetricarboxylate
Formule: C₁₀H₁₆O₆
MW: 232.23 g/mol
MDL Nummer: MFCD00009150
CAS nummer: 6279-86-3
EINECS: 228-477-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64efc3b0fde3dd163383fa8444d75d1cc23f715f&vid=64efc3b0fde3dd163383fa8444d75d1cc23f715f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR