Ammonium carbamate ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C0027-25G C0027-500G
TCIAC0027-25GEA 17.6 EUR
TCIAC0027-25G TCIAC0027-500G
Ammonium carbamate ≥97.0% (by titrimetric analysis)
Ammonium carbamate
Formule: H₂NCO₂NH₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00013010
CAS nummer: 1111-78-0
EINECS: 214-185-2
Merck Index: 14,00507
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5918c9a89a283b768f62c369c594a222a331665f&vid=5918c9a89a283b768f62c369c594a222a331665f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR