3,7-Dithia-1,9-nonanediol ≥92.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D2845-25G
D2845-5G
TCIAD2845-25GEA
588
EUR
TCIAD2845-25G
TCIAD2845-5G
3,7-Dithia-1,9-nonanediol ≥92.0% (by GC)
3,7-Dithia-1,9-nonanediol
Formule:
C₇H₁₆O₂S₂ |
MDL Nummer:
MFCD01098524 CAS nummer: 16260-48-3 |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid |
Purity (GC) | min. 92.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...