3-Thienylacetic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Thiophene-3-acetic acid, 3-Thiopheneacetic acid

T1152-25G T1152-5G
TCIAT1152-25GEA 302 EUR
TCIAT1152-25G TCIAT1152-5G
3-Thienylacetic acid ≥98.0% (by titrimetric analysis)
3-Thienylacetic acid
Formule: C₆H₆O₂S
MDL Nummer: MFCD00005473
CAS nummer: 6964-21-2
EINECS: 230-166-1
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8281f30f0bd4762485be4aeaa687c844dfa79772&vid=8281f30f0bd4762485be4aeaa687c844dfa79772&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR