5-Bromouridine ≥99.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B0666-1G B0666-100MG
TCIAB0666-1GEA 57.2 EUR
TCIAB0666-1G TCIAB0666-100MG
5-Bromouridine ≥99.0% (by HPLC)
5-Bromouridine
Formule: C₉H₁₁BrN₂O₆
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006528
CAS nummer: 957-75-5
EINECS: 213-486-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea99c00116ef91de710745cb0fdf5b7b97633dae&vid=ea99c00116ef91de710745cb0fdf5b7b97633dae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Specific rotation [a]20/D -11.0 to -14.0 deg (C=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR