Tetraethylphosphonium bromide ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T1379-25G
T1379-5G
TCIAT1379-25GEA
99
EUR
TCIAT1379-25G
TCIAT1379-5G
Tetraethylphosphonium bromide ≥98.0% (by titrimetric analysis)
Tetraethylphosphonium bromide
Formule:
C₈H₂₀BrP |
MDL Nummer:
MFCD00054230 CAS nummer: 4317-07-1 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...