5-Bromo-2-(trifluoromethyl)pyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B3451-1G B3451-5G
TCIAB3451-1GEA 45.1 EUR
TCIAB3451-1G TCIAB3451-5G
5-Bromo-2-(trifluoromethyl)pyridine ≥98.0% (by GC)
5-Bromo-2-(trifluoromethyl)pyridine
Formule: C₆H₃BrF₃N
Kookpunt: 78°C
MDL Nummer: MFCD06657686
CAS nummer: 436799-32-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=856129cd8ac5200c39bfa69d61a92f0f74d53c7a&vid=856129cd8ac5200c39bfa69d61a92f0f74d53c7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR