4-(Trifluoromethoxy)toluene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
T2454-1G T2454-5G
TCIAT2454-1GEA 38.5 EUR
TCIAT2454-1G TCIAT2454-5G
4-(Trifluoromethoxy)toluene ≥98.0% (by GC)
4-(Trifluoromethoxy)toluene
Formule: C₈H₇F₃O
MW: 176.1400 g/mol
Kookpunt: 135°C
Vlampunt: 31°C
MDL Nummer: MFCD00042413
CAS nummer: 706-27-4
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=316cc8291f0a33c2439de7b61650251c1b98720c&vid=316cc8291f0a33c2439de7b61650251c1b98720c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %