7-Ethoxycoumarin ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E0538-25G E0538-5G
TCIAE0538-25GEA 225 EUR
TCIAE0538-25G TCIAE0538-5G
7-Ethoxycoumarin ≥97.0% (by GC)
7-Ethoxycoumarin
Formule: C₁₁H₁₀O₃
MDL Nummer: MFCD00006877
CAS nummer: 31005-02-4
EINECS: 250-429-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37a722e80a0bfc2ec213e1457ef4eb20ee91131e&vid=37a722e80a0bfc2ec213e1457ef4eb20ee91131e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 88.0 to 92.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR