Isoindoline ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
I0751-1G
I0751-5G
TCIAI0751-1GEA
81.4
EUR
TCIAI0751-1G
TCIAI0751-5G
Isoindoline ≥98.0% (by titrimetric analysis)
Isoindoline
Formule:
C₈H₉N Kookpunt: 113°C Smeltpunt: 17°C Vlampunt: 94°C Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00605324 CAS nummer: 496-12-8 |
|
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...