Ethyl-4-chloroacetoacetate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Ethyl 4-chloro-3-oxobutanoate
C0911-25G C0911-500G
TCIAC0911-25GEA 17.6 EUR
TCIAC0911-25G TCIAC0911-500G
Ethyl-4-chloroacetoacetate ≥95.0% (by GC)
Ethyl-4-chloroacetoacetate
Formule: C₆H₉ClO₃
MW: 164.59 g/mol
Kookpunt: 120°C
Vlampunt: 96°C
MDL Nummer: MFCD00000939
CAS nummer: 638-07-3
EINECS: 211-317-0
UN: 2922
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86c2e71ebe13f894e5ecc4232c9a6b5018c94a71&vid=86c2e71ebe13f894e5ecc4232c9a6b5018c94a71&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %