3-Pyridylacetonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

P1232-25G P1232-5G
TCIAP1232-25GEA 277 EUR
TCIAP1232-25G TCIAP1232-5G
3-Pyridylacetonitrile ≥98.0% (by GC)
3-Pyridylacetonitrile
Formule: C₇H₆N₂
Kookpunt: 146°C
Vlampunt: 98°C
MDL Nummer: MFCD00006406
CAS nummer: 6443-85-2
EINECS: 229-241-1
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ae223e4da965a1ecdbbeca74b991885b08d79ef&vid=0ae223e4da965a1ecdbbeca74b991885b08d79ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR