Diethyl phenylphosphonate ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P0335-25ML P0335-5ML
TCIAP0335-25MLEA 258 EUR
TCIAP0335-25ML TCIAP0335-5ML
Diethyl phenylphosphonate ≥96.0% (by GC)
Diethyl phenylphosphonate
Formule: C₁₀H₁₅O₃P
Kookpunt: 267°C
MDL Nummer: MFCD00015136
CAS nummer: 1754-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f968809843d02ea08a917ba8147d1a5bd1501b7&vid=8f968809843d02ea08a917ba8147d1a5bd1501b7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %