Methyl phenylglyoxylate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B1033-25G B1033-500G B1033-100G
TCIAB1033-25GEA 33 EUR
TCIAB1033-25G TCIAB1033-500G TCIAB1033-100G
Methyl phenylglyoxylate ≥97.0% (by GC)
Methyl phenylglyoxylate
Formule: C₉H₈O₃
Kookpunt: 251°C
MDL Nummer: MFCD00008443
CAS nummer: 15206-55-0
EINECS: 239-263-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75d3efd7a2ced5923d1be72e932ca2eab2e3fae3&vid=75d3efd7a2ced5923d1be72e932ca2eab2e3fae3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR