2-Methyl-3-isopropoxypyrazine ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M0808-1G
M0808-5G
TCIAM0808-1GEA
39.6
EUR
TCIAM0808-1G
TCIAM0808-5G
2-Methyl-3-isopropoxypyrazine ≥97.0% (by GC)
2-Methyl-3-isopropoxypyrazine
Formule:
C₈H₁₂N₂O |
MDL Nummer:
MFCD00059772 CAS nummer: 94089-22-2 EINECS: 302-122-2 |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...