Diethyl ethyl(phenyl)malonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D3375-25G D3375-5G
TCIAD3375-25GEA 112 EUR
TCIAD3375-25G TCIAD3375-5G
Diethyl ethyl(phenyl)malonate ≥98.0% (by GC)
Diethyl ethyl(phenyl)malonate
Formule: C₁₅H₂₀O₄
Kookpunt: 171°C
Smeltpunt: –7°C
MDL Nummer: MFCD00040759
CAS nummer: 76-67-5
EINECS: 200-978-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b2cc23245f1e019f34f630fc022b71f921e851f&vid=1b2cc23245f1e019f34f630fc022b71f921e851f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR