Mucochloric acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M0472-25G M0472-500G
TCIAM0472-25GEA 17.6 EUR
TCIAM0472-25G TCIAM0472-500G
Mucochloric acid ≥98.0% (by GC, titration analysis)
Mucochloric acid
Formule: C₄H₂Cl₂O₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00135465
CAS nummer: 87-56-9
EINECS: 201-752-4
UN: 2923
ADR: 8,III
Merck Index: 14,06297
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d96350ed0c4051f2e0e4c814fa358a7587214d16&vid=d96350ed0c4051f2e0e4c814fa358a7587214d16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 124.0 to 128.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR