Benzohydroxamic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B0061-25G B0061-5G
TCIAB0061-25GEA 126 EUR
TCIAB0061-25G TCIAB0061-5G
Benzohydroxamic acid ≥98.0% (by titrimetric analysis)
Benzohydroxamic acid
Formule: C₇H₇NO₂
MDL Nummer: MFCD00002109
CAS nummer: 495-18-1
EINECS: 207-797-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bd7b9cbc13c13ab54a5ed8e6470d031b5ea87b7&vid=9bd7b9cbc13c13ab54a5ed8e6470d031b5ea87b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR